Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144527.3 | NP_653110.3 | 4 | 591 | ||
GenBank mRNA | AK030742.1 | - | 4 | 556 | ||
AK085545.1 | - | 4 | 538 | |||
AK141103.1 | - | 4 | 581 | |||
BC008221.1 | - | 4 | 543 | |||
BC031729.1 | - | 4 | 487 | 3992639 |