Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kazn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109684.1 | NP_001103154.1 | 7 | 1285 | ||
NM_001109685.1 | NP_001103155.1 | 8 | 1303 | |||
NM_144531.3 | NP_653114.3 | 7 | 1285 | |||
GenBank mRNA | AK018497.1 | - | 8 | 1283 | ||
AK032460.1 | - | 5 | 524 | |||
AK052809.1 | - | 8 | 1304 | |||
AK173090.1 | - | 7 | 1285 | |||
BC022941.1 | - | 7 | 1289 | |||
BC100387.1 | - | 5 | 505 |