Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgi4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144556.2 | NP_653139.2 | 1 | 199 | ||
GenBank mRNA | AJ487521.1 | - | 1 | 200 | ||
AJ487960.1 | - | 1 | 199 | |||
AK033967.1 | - | 1 | 17 | |||
AK134643.1 | - | 1 | 793 | |||
AK148793.1 | - | 1 | 458 | |||
BC098469.1 | - | 1 | 358 | 30099294 |