Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144783.2 | NP_659032.3 | 8 | 1568 | ||
GenBank mRNA | BG087452.2 | - | 4 | 367 | ||
DQ537939.1 | - | 8 | 905 | |||
M55512.1 | - | 8 | 1573 |