Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000627.3 | NP_000618.3 | 14, 15 | 2162 | ||
NM_001166264.1 | NP_001159736.1 | 14, 15 | 2162 | |||
NM_001166265.1 | NP_001159737.1 | 14, 15 | 2003 | |||
NM_001166266.1 | NP_001159738.1 | 14, 15 | 2003 | |||
NM_206943.2 | NP_996826.2 | 18, 19 | 3022 | |||
XM_005264317.3 | XP_005264374.1 | 18, 19 | 2865 | |||
XM_005264318.3 | XP_005264375.1 | 18, 19 | 2865 | |||
XM_011532853.2 | XP_011531155.1 | 18, 19 | 3198 | |||
XM_011532855.2 | XP_011531157.1 | 18, 19 | 3021 | |||
XM_011532857.2 | XP_011531159.1 | 18, 19 | 3027 | |||
XM_011532858.2 | XP_011531160.1 | 17, 18 | 2901 | |||
XM_011532859.2 | XP_011531161.1 | 18, 19 | 2868 | |||
XM_011532860.2 | XP_011531162.1 | 17, 18 | 2859 | |||
XM_011532861.2 | XP_011531163.1 | 18, 19 | 3027 | |||
XM_011532862.2 | XP_011531164.1 | 18, 19 | 3027 | |||
XM_017004109.1 | XP_016859598.1 | 18, 19 | 3024 | |||
GenBank mRNA | AB208801.1 | - | 14, 15 | 1961 | ||
AF489528.1 | - | 14, 15 | 2018 | |||
AK298027.1 | - | 8, 9 | 731 | |||
AK299912.1 | - | 9, 10 | 960 | |||
BC130289.1 | - | 14, 15 | 2102 | 40146320 | ||
BC144127.1 | - | 14, 15 | 1945 | |||
BC144128.1 | - | 14, 15 | 2102 | 9052648 | ||
M34057.1 | - | 14, 15 | 2108 |