Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172095.1 | NP_001165566.1 | 2, 3 | 302 | ||
NM_144787.2 | NP_659036.1 | 2, 3 | 366 | |||
GenBank mRNA | AK048092.1 | - | 2, 3 | 279 | ||
AK053104.1 | - | 2, 3 | 242 | |||
AK081769.1 | - | 2, 3 | 309 | |||
AK144103.1 | - | 2, 3 | 329 | |||
AK144890.1 | - | 2, 3 | 340 | |||
AK173023.1 | - | 2, 3 | 366 | |||
BC020180.1 | - | 2, 3 | 308 | 4923943 | ||
BC042424.1 | - | 2, 3 | 302 | 5357211 |