Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144799.2 | NP_659048.1 | 3 | 468 | ||
GenBank mRNA | AK075847.1 | - | 3 | 469 | ||
AK159946.1 | - | 3 | 419 | |||
BC019124.1 | - | 3 | 419 | 5007525 |