Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reep6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204931.1 | NP_001191860.1 | 5 | 1644 | ||
NM_139292.2 | NP_647453.1 | 6 | 1725 | |||
GenBank mRNA | AB039933.1 | - | 6 | 1700 | ||
AK002562.1 | - | 5 | 1637 | |||
BC029741.1 | - | 5 | 1631 | 4195889 |