Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il31ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139299.2 | NP_647460.2 | 17 | 2332 | ||
GenBank mRNA | AB083111.1 | - | 17 | 2331 | ||
AF486621.1 | - | 14 | 1911 | |||
AK030512.1 | - | 6 | 901 | |||
BC145083.1 | - | 17 | 2341 | |||
BC145084.1 | - | 16 | 2235 | |||
BC145085.1 | - | 17 | 2358 |