Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139305.2 | NP_647466.2 | 11 | 1356 | ||
GenBank mRNA | AB086322.1 | - | 9 | 1060 | ||
AJ245857.1 | - | 11 | 1358 | |||
AK136579.1 | - | 11 | 1357 | |||
AK137719.1 | - | 11 | 1358 | |||
AK166403.1 | - | 8 | 785 | |||
BC120544.1 | - | 11 | 1349 | 40129467 | ||
BC120546.1 | - | 11 | 1349 | 40129469 | ||
EF122497.1 | - | 9 | 1064 |