Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144500.3 | NP_653083.2 | 9 | 1062 | ||
GenBank mRNA | AK048537.1 | - | 9 | 982 | ||
AK145292.1 | - | 9 | 1094 | |||
AK161695.1 | - | 9 | 1001 | |||
AK166059.1 | - | 9 | 1041 | |||
AK169504.1 | - | 2 | 987 | |||
AK170306.1 | - | 9 | 1046 | |||
AK170669.1 | - | 9 | 1063 | |||
AK178638.1 | - | 3 | 241 | |||
AK182769.1 | - | 3 | 250 | |||
AK220257.1 | - | 3 | 381 | |||
BC026804.1 | - | 9 | 1069 | 4159217 |