Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144537.3 | NP_653120.1 | 9, 10 | 1376 | ||
GenBank mRNA | AF466283.1 | - | 9, 10 | 1377 | ||
AK015256.1 | - | 9, 10 | 1379 | |||
BC138114.1 | - | 8, 9 | 894 | 8861133 |