Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3il1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033756.1 | - | 1 | 1860 | ||
AK085375.1 | - | 1 | 2848 | |||
BC020147.1 | - | 11 | 1938 | 3708973 |