Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slamf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144539.5 | NP_653122.2 | 8 | 2402 | ||
GenBank mRNA | AK030135.1 | - | 8 | 2263 | ||
AK030148.1 | - | 8 | 2403 |