Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000641.3 | NP_000632.1 | 2, 3 | 343 | ||
GenBank mRNA | AK290572.1 | - | 2, 3 | 343 | ||
AK315294.1 | - | 2, 3 | 343 | |||
BC012506.1 | - | 2, 3 | 326 | 4476678 | ||
M57765.1 | - | 2, 3 | 326 | |||
X58377.1 | - | 2, 3 | 253 |