Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trib2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144551.5 | NP_653134.2 | 2, 3 | 1845 | ||
GenBank mRNA | AF358867.1 | - | 2, 3 | 710 | ||
AK044747.1 | - | 2, 3 | 1803 | |||
AK080064.1 | - | 2, 3 | 1048 | |||
AK082329.1 | - | 2, 3 | 1057 | |||
AK144192.1 | - | 2, 3 | 1910 | |||
BC027159.1 | - | 1, 2 | 302 | |||
BC034338.1 | - | 2, 3 | 879 | 5043179 | ||
BC037387.1 | - | 2, 3 | 1789 | 5363174 |