Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000573.3 | NP_000564.2 | 2 | 306 | ||
NM_000651.4 | NP_000642.3 | 2 | 306 | |||
XM_006711166.2 | XP_006711229.1 | 2 | 282 | |||
XM_011509205.1 | XP_011507507.1 | 2 | 282 | |||
GenBank mRNA | AK298486.1 | - | 2 | 197 | ||
AK309342.1 | - | 2 | 287 | |||
BC032550.1 | - | 2 | 195 | |||
X14362.1 | - | 2 | 93 | |||
Y00816.1 | - | 193 |
Gene Symbol: CR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175710.1 | NP_783641.1 | 2 | 263 | ||
GenBank mRNA | BC109190.1 | - | 2 | 202 | ||
BC109191.1 | - | 2 | 202 | |||
BX094042.1 | - | 2 | 263 |