Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146690.1 | NP_001140162.1 | 1, 2 | 394 | ||
NM_144807.3 | NP_659056.3 | 1, 2 | 394 | |||
GenBank mRNA | AK007964.1 | - | 1, 2 | 314 | ||
AK032497.1 | - | 1, 2 | 289 | |||
AY445814.1 | - | 1, 2 | 289 | |||
BC016251.1 | - | 1, 2 | 342 | 4911104 | ||
BC031435.1 | - | 1, 2 | 349 | 4221062 | ||
BY341208.1 | - | 1, 2 | 302 |