Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135151.1 | NP_001128623.1 | 4 | 674 | ||
NM_001135152.1 | NP_001128624.1 | 4 | 640 | |||
NM_144808.4 | NP_659057.2 | 4 | 674 | |||
GenBank mRNA | AB177995.1 | - | 4 | 677 | ||
AK090193.1 | - | 4 | 639 | |||
AK122197.1 | - | 2 | 291 | |||
AK158096.1 | - | 2 | 2603 | |||
BC021530.1 | - | 4 | 677 | 5353717 |