Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252582.1 | NP_001239511.1 | 3 | 585 | ||
NM_001252583.1 | NP_001239512.1 | 3 | 550 | |||
NM_144833.3 | NP_659082.1 | 3 | 550 | |||
GenBank mRNA | AF295806.1 | - | 3 | 312 | ||
AK033090.1 | - | 3 | 327 | |||
AK042224.1 | - | 3 | 307 | |||
AK049384.1 | - | 4 | 605 | |||
AK053001.1 | - | 3 | 275 | |||
AK083003.1 | - | 3 | 292 | |||
AK132345.1 | - | 3 | 288 | |||
AK139797.1 | - | 2 | 104 | |||
BC021528.1 | - | 3 | 305 | 5353673 | ||
BC052080.1 | - | 3 | 282 | 6403411 | ||
BC056957.1 | - | 3 | 279 | |||
CK791111.1 | - | 3 | 550 |