Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k7cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144854.2 | NP_659103.1 | 4 | 484 | ||
GenBank mRNA | AK041664.1 | - | 4 | 394 | ||
AY033899.1 | - | 4 | 499 | |||
BC115624.1 | - | 4 | 258 | 40102836 | ||
BC115625.1 | - | 4 | 258 | 40102837 | ||
BX519631.1 | - | 4 | 485 |