Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000669.4 | NP_000660.1 | 3 | 511 | ||
NR_133005.1 | 3 | 511 | ||||
XM_011531588.2 | XP_011529890.1 | 2 | 301 | |||
XM_011531589.1 | XP_011529891.1 | 4 | 631 | |||
GenBank mRNA | BC062476.1 | - | 3 | 207 | 30408761 | |
BC066227.1 | - | 3 | 169 | 7002195 | ||
BC066228.1 | - | 3 | 169 | 7002197 | ||
BC067419.1 | - | 3 | 169 | 7002191 | ||
BC067420.1 | - | 3 | 169 | 7002192 | ||
BC067421.1 | - | 3 | 169 | 7002193 | ||
BC067422.1 | - | 3 | 169 | 7002194 | ||
BC074771.2 | - | 3 | 169 | 30915457 | ||
BC074786.2 | - | 3 | 168 | 30915344 | ||
CB114902.1 | - | 3 | 226 | |||
CD014078.1 | - | 3 | 212 | |||
M12272.1 | - | 3 | 221 | |||
X04299.1 | - | 3 | 213 |