Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110015.1 | NP_001103485.1 | 3 | 387 | ||
NM_001110016.1 | NP_001103486.1 | 3 | 387 | |||
NM_144863.4 | NP_659112.2 | 3 | 387 | |||
GenBank mRNA | AK041763.1 | - | 3 | 307 | ||
AK045396.1 | - | 3 | 360 | |||
AK077545.1 | - | 3 | 317 | |||
AK140019.1 | - | 3 | 312 | |||
AK155965.1 | - | 3 | 286 | |||
AK156822.1 | - | 388 | ||||
AK171683.1 | - | 3 | 325 | |||
AK172054.1 | - | 3 | 312 | |||
AK172377.1 | - | 3 | 317 | |||
BC022132.1 | - | 3 | 359 | |||
BC025809.1 | - | 3 | 301 | |||
BC094453.1 | - | 3 | 338 | 5068941 |