Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039482.1 | NP_001034571.1 | 3 | 571 | ||
GenBank mRNA | AK032131.1 | - | 1 | 58 | ||
AK053102.1 | - | 3 | 521 | |||
AK053371.1 | - | 3 | 535 | |||
AK054169.1 | - | 3 | 539 | |||
AK220547.1 | - | 3 | 550 | |||
BC019571.1 | - | 3 | 543 | 4235202 | ||
BC060183.1 | - | 3 | 500 |