Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144896.4 | NP_659145.1 | 1, 2 | 193 | ||
GenBank mRNA | AK028193.1 | - | 1, 2 | 192 | ||
AK034702.1 | - | 1, 2 | 194 | |||
AK053978.1 | - | 1, 2 | 196 | |||
AK081800.1 | - | 1, 2 | 197 | |||
AK134831.1 | - | 1, 2 | 128 | |||
AK166231.1 | - | 1, 2 | 204 | |||
BC005709.1 | - | 1, 2 | 195 | 3157973 |