Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161854.1 | NP_001155326.1 | 4 | 789 | ||
NM_144901.3 | NP_659150.1 | 4 | 789 | |||
GenBank mRNA | AK044837.1 | - | 2 | 1407 | ||
AK122393.1 | - | 4 | 699 | |||
AK144097.1 | - | 4 | 729 | |||
AK145351.1 | - | 4 | 894 | |||
AK145428.1 | - | 4 | 711 | |||
AK145932.1 | - | 4 | 706 | |||
AK146586.1 | - | 4 | 728 | |||
AK146706.1 | - | 4 | 721 | |||
AK147012.1 | - | 4 | 728 | |||
AK150882.1 | - | 4 | 706 | |||
AK151638.1 | - | 4 | 725 | |||
AK151723.1 | - | 4 | 729 | |||
AK151970.1 | - | 4 | 729 | |||
AK152531.1 | - | 4 | 725 | |||
AK153196.1 | - | 4 | 706 | |||
AK154068.1 | - | 4 | 714 | |||
AK155751.1 | - | 4 | 714 | |||
AK159258.1 | - | 4 | 722 | |||
AK159863.1 | - | 4 | 722 | |||
AK168556.1 | - | 3 | 523 | |||
BC016898.1 | - | 3 | 633 | 4224466 | ||
BC029688.1 | - | 4 | 725 | 5367783 | ||
BC031404.1 | - | 4 | 697 | 4953772 | ||
BC062097.1 | - | 4 | 704 | 6833505 |