Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168256.1 | NP_001161728.1 | 8 | 621 | ||
NM_001168257.1 | NP_001161729.1 | 7 | 546 | |||
NM_001168258.1 | NP_001161730.1 | 3 | 346 | |||
NM_001168259.1 | NP_001161731.1 | 4 | 113 | |||
NM_144805.2 | NP_659054.2 | 8 | 489 | |||
GenBank mRNA | AK078864.1 | - | 4 | 114 | ||
BC019416.1 | - | 8 | 451 | 5151159 | ||
BX511925.1 | - | 3 | 73 | |||
CT010303.1 | - | 7 | 419 | |||
CV561368.1 | - | 7 | 488 | |||
CX564997.1 | - | 8 | 520 |