Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpsap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164242.1 | NP_001157714.1 | 3, 4 | 334 | ||
NM_144806.2 | NP_659055.1 | 4, 5 | 430 | |||
GenBank mRNA | AK038680.1 | - | 4, 5 | 396 | ||
AK076811.1 | - | 4, 5 | 384 | |||
AK077330.1 | - | 4, 5 | 383 | |||
BC023178.1 | - | 4, 5 | 415 | 4947226 | ||
BC106185.1 | - | 3, 4 | 263 | 6512229 |