Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144816.1 | NP_659065.1 | 3 | 330 | ||
GenBank mRNA | AK140428.1 | - | 3 | 361 | ||
BC021549.1 | - | 3 | 330 | 4501997 |