Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem106a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144830.3 | NP_659079.1 | 7 | 781 | ||
GenBank mRNA | AK136954.1 | - | 3 | 1775 | ||
AK146019.1 | - | 7 | 782 | |||
AK149650.1 | - | 6 | 1025 | |||
AK150285.1 | - | 7 | 751 | |||
AK151842.1 | - | 7 | 757 | |||
AK152741.1 | - | 7 | 751 | |||
AK154217.1 | - | 7 | 782 | |||
AK168437.1 | - | 7 | 725 | |||
AK169081.1 | - | 7 | 724 | |||
AK170900.1 | - | 7 | 752 | |||
AK172282.1 | - | 7 | 730 | |||
BC022145.1 | - | 6 | 829 | 5101508 | ||
BC033268.1 | - | 7 | 754 | 5037750 |