Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000689.4 | NP_000680.2 | 2 | 478 | ||
GenBank mRNA | AB209821.1 | - | 2 | 204 | ||
AF003341.1 | - | 2 | 161 | |||
AK000118.1 | - | 2 | 205 | |||
AK026641.1 | - | 2 | 205 | |||
AK293176.1 | - | 2 | 171 | |||
AK302168.1 | - | 2 | 214 | |||
AY390731.1 | - | 2 | 161 | |||
BC001505.2 | - | 2 | 186 | 2988388 | ||
BG773210.1 | - | 2 | 220 | |||
BT006921.1 | - | 2 | 161 | |||
DC398835.1 | - | 2 | 497 | |||
EU668318.1 | - | 2 | 205 | |||
EU794589.1 | - | 2 | 161 | |||
FJ224286.1 | - | 2 | 214 |