Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144847.1 | NP_659096.1 | 7, 8 | 643 | ||
GenBank mRNA | BC011468.1 | - | 7, 8 | 576 | 4012880 | |
BC012437.1 | - | 7, 8 | 643 | 3710089 | ||
BC025042.1 | - | 2, 3 | 105 | |||
BC068117.1 | - | 12, 13 | 1291 |