Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pja2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025309.1 | NP_001020480.1 | 3 | 284 | ||
NM_144859.2 | NP_659108.1 | 3 | 284 | |||
GenBank mRNA | AF493070.1 | - | 3 | 341 | ||
AK122282.1 | - | 3 | 243 | |||
AK144586.1 | - | 3 | 271 | |||
AK164025.1 | - | 3 | 289 | |||
AK168371.1 | - | 3 | 266 | |||
BC017130.1 | - | 2 | 210 | 4506259 | ||
BY065517.1 | - | 3 | 294 |