Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252573.1 | NP_001239502.1 | 9, 10 | 1295 | ||
NM_001252574.1 | NP_001239503.1 | 9, 10 | 1230 | |||
NM_001252575.1 | NP_001239504.1 | 9, 10 | 1178 | |||
NM_144893.2 | NP_659142.1 | 9, 10 | 1249 | |||
GenBank mRNA | AK053180.1 | - | 9, 10 | 1165 | ||
AK155539.1 | - | 9, 10 | 1264 | |||
AK160899.1 | - | 9, 10 | 1235 | |||
AK170715.1 | - | 9, 10 | 1200 | |||
BC018327.1 | - | 10, 11 | 1115 | 4166249 | ||
BC064092.1 | - | 1, 2 | 382 | |||
BC094025.1 | - | 9, 10 | 1228 | 4488690 |