Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH9A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000696.3 | NP_000687.3 | 2 | 324 | ||
XM_011509294.2 | XP_011507596.1 | 2 | 388 | |||
GenBank mRNA | AF172093.1 | - | 2 | 147 | ||
AK293520.1 | - | 2 | 228 | |||
AK301767.1 | - | 2 | 189 | |||
AK302183.1 | - | 2 | 155 | |||
AK302191.1 | - | 2 | 97 | |||
AK302654.1 | - | 2 | 375 | |||
AK312751.1 | - | 2 | 253 | |||
BC070030.1 | - | 2 | 253 | |||
BC151140.1 | - | 2 | 312 | 9057040 | ||
BC151141.1 | - | 2 | 312 | 9057041 | ||
U34252.1 | - | 2 | 521 | |||
U50203.1 | - | 2 | 63 | |||
X75425.1 | - | 1 | 51 |