Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144898.2 | NP_659147.2 | 2, 3 | 302 | ||
GenBank mRNA | AK163280.1 | - | 2, 3 | 162 | ||
AK167586.1 | - | 2, 3 | 250 | |||
AK170160.1 | - | 2, 3 | 285 | |||
BC008103.1 | - | 2, 3 | 252 | 3966815 | ||
BY267786.1 | - | 2, 3 | 303 |