Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144901.3 | NP_659150.1 | 5 | 932 | ||
GenBank mRNA | AK044837.1 | - | 2 | 1949 | ||
AK146706.1 | - | 5 | 864 | |||
AK155751.1 | - | 5 | 857 | |||
BC016898.1 | - | 4 | 776 | 4224466 | ||
BC062097.1 | - | 5 | 847 | 6833505 |