Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301404.1 | NP_001288333.1 | 2 | 630 | ||
NM_144834.4 | NP_659083.2 | 2 | 630 | |||
GenBank mRNA | AK050552.1 | - | 607 | |||
AY324633.1 | - | 2 | 630 | |||
BC018416.1 | - | 2 | 607 | 4196269 | ||
BC025821.1 | - | 1 | 592 | 5100400 |