Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144835.4 | NP_659084.3 | 41 | 5928 | ||
GenBank mRNA | AK032134.1 | - | 6 | 853 | ||
AK132192.1 | - | 1602 | ||||
BC019693.1 | - | 5 | 567 | |||
BC066225.1 | - | 1 | 1236 | |||
BC069833.1 | - | 1 | 1236 |