Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144843.4 | NP_659092.1 | 14 | 3119 | ||
GenBank mRNA | AK076218.1 | - | 14 | 3097 | ||
AK154859.1 | - | 14 | 3039 | |||
BC020019.1 | - | 14 | 3053 | 3596732 | ||
BC022979.1 | - | 4 | 2160 | |||
CB520796.1 | - | 1 | 576 |