Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam49b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144846.5 | NP_659095.1 | 12 | 1829 | ||
GenBank mRNA | AK028062.1 | - | 12 | 1830 | ||
AK075613.1 | - | 13 | 1774 | |||
BC011343.1 | - | 12 | 1646 | 4012217 | ||
CK633763.1 | - | 1 | 101 |