Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177810.1 | NP_001171281.1 | 28 | 3617 | ||
NM_001177811.1 | NP_001171282.1 | 27 | 3566 | |||
NM_144850.2 | NP_659099.2 | 28 | 3593 | |||
GenBank mRNA | AK034265.1 | - | 15 | 1957 | ||
AK142594.1 | - | 28 | 3511 | |||
AK162003.1 | - | 11 | 1540 | |||
AK169430.1 | - | 27 | 3565 | |||
BC020311.2 | - | 10 | 1435 | |||
BC020532.1 | - | 28 | 3605 | 4236136 |