Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k7cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144854.2 | NP_659103.1 | 5 | 811 | ||
GenBank mRNA | AK041664.1 | - | 5 | 721 | ||
AY033899.1 | - | 5 | 826 | |||
BC115624.1 | - | 5 | 585 | 40102836 | ||
BC115625.1 | - | 5 | 585 | 40102837 |