Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uhrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144873.2 | NP_659122.2 | 14, 15 | 2435 | ||
GenBank mRNA | AB116653.1 | - | 14, 15 | 2170 | ||
AF274047.1 | - | 14, 15 | 2345 | |||
AK075912.1 | - | 3, 4 | 380 | |||
AK170648.1 | - | 5, 6 | 591 | |||
BC060241.1 | - | 14, 15 | 2435 | 6334856 |