Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144874.4 | NP_659123.2 | 9 | 1890 | ||
GenBank mRNA | AK048099.1 | - | 9 | 1872 | ||
AK051792.1 | - | 9 | 1789 | |||
AK141631.1 | - | 9 | 1822 | |||
AK201289.1 | - | 1 | 233 | |||
BC021498.1 | - | 9 | 1810 |