Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vash2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144879.2 | NP_659128.2 | 8 | 3022 | ||
GenBank mRNA | AK045261.1 | - | 8 | 2818 | ||
AK140989.1 | - | 8 | 3016 | |||
AK163193.1 | - | 8 | 2798 | |||
BC024141.1 | - | 8 | 2792 | 5149750 |