Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spg20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144987.1 | NP_001138459.1 | 9 | 2335 | ||
NM_001144988.1 | NP_001138460.1 | 8 | 1948 | |||
NM_144895.2 | NP_659144.1 | 9 | 2119 | |||
GenBank mRNA | AK031545.1 | - | 8 | 1674 | ||
AK035081.1 | - | 7 | 1807 | |||
AK036569.1 | - | 7 | 1271 | |||
AK087388.1 | - | 9 | 2088 | |||
AK129171.1 | - | 8 | 1927 | |||
AK147760.1 | - | 9 | 2336 | |||
AK159951.1 | - | 9 | 2120 | |||
AK193708.1 | - | 2 | 299 | |||
AK201628.1 | - | 2 | 298 | |||
BC022921.1 | - | 9 | 2087 | 4220506 | ||
BC026683.1 | - | 1 | 281 |