Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287218.1 | NP_001274147.1 | 2, 3 | 309 | ||
NM_001287220.1 | NP_001274149.1 | 2, 3 | 309 | |||
NM_012424.4 | NP_036556.2 | 2, 3 | 309 | |||
XM_005273095.1 | XP_005273152.1 | 2, 3 | 296 | |||
XM_006711272.1 | XP_006711335.1 | 2, 3 | 296 | |||
XM_017001025.1 | XP_016856514.1 | 2, 3 | 282 | |||
XM_017001026.1 | XP_016856515.1 | 2, 3 | 284 | |||
XM_017001032.1 | XP_016856521.1 | 2, 3 | 284 | |||
GenBank mRNA | AB070706.1 | - | 2, 3 | 137 | ||
AF477978.1 | - | 2, 3 | 286 | |||
AK122989.1 | - | 2, 3 | 296 | |||
BC104769.1 | - | 2, 3 | 223 | 8143772 | ||
BC143473.1 | - | 2, 3 | 223 | |||
DB060692.1 | - | 2, 3 | 297 |