Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4BPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000716.3 | NP_000707.1 | 2 | 412 | ||
NM_001017364.1 | NP_001017364.1 | 2 | 409 | |||
NM_001017365.2 | NP_001017365.1 | 3 | 274 | |||
NM_001017366.2 | NP_001017366.1 | 3 | 271 | |||
NM_001017367.1 | NP_001017367.1 | 3 | 248 | |||
XM_005273254.4 | XP_005273311.1 | 4 | 553 | |||
XM_005273255.2 | XP_005273312.1 | 2 | 333 | |||
GenBank mRNA | AK293379.1 | - | 2 | 114 | ||
AK315137.1 | - | 2 | 333 | |||
BC005378.1 | - | 2 | 409 | 3934453 | ||
BG568018.1 | - | 3 | 273 | |||
L11245.1 | - | 2 | 397 | |||
M29964.1 | - | 2 | 147 |