Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144922.2 | NP_659171.1 | 3 | 614 | ||
NM_178089.2 | NP_835190.1 | 3 | 595 | |||
GenBank mRNA | AK042090.1 | - | 3 | 595 | ||
AK087545.1 | - | 3 | 584 | |||
AK143666.1 | - | 3 | 657 | |||
AK155604.1 | - | 3 | 625 | |||
AK202662.1 | - | 2 | 247 | |||
AK205010.1 | - | 2 | 247 | |||
BC021506.1 | - | 3 | 638 | 5347099 | ||
BC078642.1 | - | 3 | 538 | 6329630 |